Assay Details
| Gene Symbol: Als2cr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175200.4 | NP_780409.2 | 2 - 3 | 494 | 99 | |
| GenBank mRNA | AK076617.1 | - | 2 - 3 | 496 | 99 | |
| BB614746.1 | - | 2 - 3 | 495 | 99 | ||
| BC118929.2 | - | 2 - 3 | 460 | 40090362 | 99 | |
More Information
Set Membership: |
|

Als2cr11
Genomic Map