Assay Details
| Gene Symbol: Fam78b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175461.4 | NP_780670.2 | 2 - 3 | 1347 | 71 | |
| GenBank mRNA | AK033840.1 | - | 1 - 2 | 845 | 71 | |
| BC066094.1 | - | 2 - 3 | 1175 | 71 | ||
More Information
Set Membership: |
|

Fam78b
Genomic Map