Assay Details
| Gene Symbol: Dcaf12l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175539.3 | NP_780748.1 | 1 | 1614 | 89 | |
| GenBank mRNA | AK037333.1 | - | 2 | 1564 | 89 | |
| AK076436.1 | - | 1621 | 89 | |||
More Information
Set Membership: |
|

Dcaf12l2
Genomic Map