Assay Details
| Gene Symbol: Ush1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176847.3 | NP_789817.1 | 2 - 3 | 1441 | 68 | |
| GenBank mRNA | AB087502.1 | - | 2 - 3 | 1441 | 68 | |
| BC120509.1 | - | 2 - 3 | 1424 | 40129432 | 68 | |
| BC137807.1 | - | 2 - 3 | 1424 | 8860826 | 68 | |
More Information
Set Membership: |
|

Ush1g
Genomic Map