Assay Details
Gene Symbol: Bhlha9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177182.4 | NP_796156.3 | 1 | 724 | 60 | |
GenBank mRNA | AK043927.1 | - | 1 | 469 | 60 | |
BC048728.1 | - | 1 | 724 | 6510150 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
