Assay Details
| Gene Symbol: Spata33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177279.4 | NP_796253.2 | 1 - 2 | 142 | 97 | |
| GenBank mRNA | AK076656.1 | - | 1 - 2 | 118 | 97 | |
More Information
Set Membership: |
|

Spata33
Genomic Map