Assay Details
| Gene Symbol: Zfp771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177362.3 | NP_796336.1 | 2 - 3 | 236 | 114 | |
| GenBank mRNA | AK090240.1 | - | 2 - 3 | 237 | 114 | |
| AK159461.1 | - | 2 - 3 | 231 | 114 | ||
| BC139048.1 | - | 2 - 3 | 219 | 8862070 | 114 | |
| BC139049.1 | - | 2 - 3 | 219 | 8862071 | 114 | |
| BC145524.1 | - | 2 - 3 | 232 | 9054087 | 114 | |
More Information
Set Membership: |
|

Zfp771
Genomic Map