Assay Details
| Gene Symbol: Gemin4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177367.3 | NP_796341.2 | 1 - 2 | 96 | 59 | |
| GenBank mRNA | AK133821.1 | - | 1 - 2 | 188 | 59 | |
| AK154061.1 | - | 1 - 2 | 77 | 59 | ||
| AK165864.1 | - | 1 - 2 | 40 | 59 | ||
| BC031425.1 | - | 1 - 2 | 36 | 3982927 | 59 | |
| BC062157.1 | - | 1 - 2 | 44 | 6508223 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK154061.1, BC031425.1, BC062157.1, NM_177367.3 transcripts.
Set Membership: |
|

Gemin4
Genomic Map