Assay Details
| Gene Symbol: Lcmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177846.3 | NP_808514.2 | 1 | 1923 | 89 | |
| GenBank mRNA | AK038626.1 | - | 1604 | 89 | ||
| AK054392.1 | - | 1 | 1613 | 89 | ||
| AK084642.1 | - | 1 | 1891 | 89 | ||
| AK172552.1 | - | 1 | 1604 | 89 | ||
| AK172970.1 | - | 1 | 1878 | 89 | ||
| BC071272.1 | - | 1 | 1866 | 89 | ||
More Information
Set Membership: |
|

Lcmt2
Genomic Map