Assay Details
| Gene Symbol: Zfp866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177899.3 | NP_808567.2 | 2 - 3 | 251 | 69 | |
| GenBank mRNA | AK052363.1 | - | 2 - 3 | 230 | 69 | |
| AK137334.1 | - | 2 - 3 | 253 | 69 | ||
| AK140357.1 | - | 2 - 3 | 246 | 69 | ||
| BC057095.1 | - | 2 - 3 | 225 | 6419770 | 69 | |
| BC117846.1 | - | 2 - 3 | 206 | 40093653 | 69 | |
| BC117847.1 | - | 2 - 3 | 206 | 40093655 | 69 | |
| BC138224.1 | - | 2 - 3 | 202 | 8861243 | 69 | |
| BC138227.1 | - | 2 - 3 | 202 | 8861246 | 69 | |
More Information
Set Membership: |
|

Zfp866
Genomic Map