Assay Details
| Gene Symbol: Hist1h2ac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178189.5 | NP_835496.1 | 1 | 421 | 65 | |
| GenBank mRNA | AK033518.1 | - | 423 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h2ac| Gene Symbol: Hist1h2ac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178189.5 | NP_835496.1 | 1 | 421 | 65 | |
| GenBank mRNA | AK033518.1 | - | 423 | 65 | ||
Set Membership: |
|