Assay Details
| Gene Symbol: Egfl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164564.1 | NP_001158036.1 | 2 - 3 | 182 | 67 | |
| NM_178444.4 | NP_848538.3 | 3 - 4 | 407 | 67 | ||
| NM_198724.2 | NP_942017.2 | 3 - 4 | 406 | 67 | ||
| NM_198725.2 | NP_942018.2 | 3 - 4 | 406 | 67 | ||
| GenBank mRNA | AF184973.1 | - | 3 - 4 | 357 | 67 | |
| AK002601.1 | - | 3 - 4 | 402 | 67 | ||
| AK139377.1 | - | 2 - 3 | 177 | 67 | ||
| AK156158.1 | - | 3 - 4 | 408 | 67 | ||
| AY239289.1 | - | 3 - 4 | 368 | 67 | ||
| AY239290.1 | - | 3 - 4 | 390 | 67 | ||
| AY309459.1 | - | 3 - 4 | 380 | 67 | ||
| BB871615.1 | - | 2 - 3 | 183 | 67 | ||
| BC024610.1 | - | 3 - 4 | 390 | 4159675 | 67 | |
| BY274501.1 | - | 3 - 4 | 406 | 67 | ||
More Information
Set Membership: |
|

Egfl7
Genomic Map