Assay Details
| Gene Symbol: Egfl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164564.1 | NP_001158036.1 | 8 - 9 | 896 | 85 | |
| NM_178444.4 | NP_848538.3 | 9 - 10 | 1121 | 85 | ||
| NM_198724.2 | NP_942017.2 | 9 - 10 | 1120 | 85 | ||
| GenBank mRNA | AF184973.1 | - | 9 - 10 | 1071 | 85 | |
| AK139377.1 | - | 8 - 9 | 891 | 85 | ||
| AK156158.1 | - | 9 - 10 | 1123 | 85 | ||
| AY239289.1 | - | 9 - 10 | 1082 | 85 | ||
| AY239290.1 | - | 9 - 10 | 1104 | 85 | ||
| AY309459.1 | - | 9 - 10 | 1094 | 85 | ||
| BC024610.1 | - | 9 - 10 | 1104 | 4159675 | 85 | |
| CF949270.1 | - | 1 - 2 | 157 | 85 | ||
More Information
Set Membership: |
|

Egfl7
Genomic Map