Assay Details
| Gene Symbol: Yod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178691.4 | NP_848806.2 | 1 - 2 | 487 | 78 | |
| GenBank mRNA | AK036938.1 | - | 1 - 2 | 488 | 78 | |
| AK036991.1 | - | 1 - 2 | 489 | 78 | ||
| AK044678.1 | - | 1 - 2 | 282 | 78 | ||
| AK053730.1 | - | 1 - 2 | 277 | 78 | ||
| BC099948.1 | - | 1 - 2 | 280 | 30097649 | 78 | |
| BC139034.1 | - | 1 - 2 | 413 | 8862056 | 78 | |
More Information
Set Membership: |
|

Yod1
Genomic Map