Assay Details
| Gene Symbol: Adm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182928.4 | NP_891558.1 | 1 - 2 | 232 | 109 | |
| GenBank mRNA | AB121035.1 | - | 1 - 2 | 156 | 109 | |
| BC096428.1 | - | 1 - 2 | 159 | 4948766 | 109 | |
| CJ052708.1 | - | 1 - 2 | 233 | 109 | ||
| DV649647.1 | - | 1 - 2 | 108 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB121035.1, BC096428.1, NM_182928.4 transcripts.
Set Membership: |
|

Adm2
Genomic Map