Assay Details
| Gene Symbol: Ap1s3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183027.2 | NP_898848.1 | 1 - 2 | 123 | 97 | |
| GenBank mRNA | AK028013.1 | - | 1 - 2 | 108 | 97 | |
| BC054111.1 | - | 1 - 2 | 127 | 3154405 | 97 | |
| BC098237.1 | - | 1 - 2 | 132 | 3825593 | 97 | |
| BF715142.1 | - | 1 - 2 | 123 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC054111.1, BC098237.1, NM_183027.2 transcripts.
Set Membership: |
|

Ap1s3
Genomic Map