Assay Details
| Gene Symbol: Ap1s3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183027.2 | NP_898848.1 | 2 - 3 | 305 | 95 | |
| GenBank mRNA | AK028013.1 | - | 2 - 3 | 290 | 95 | |
| BC054111.1 | - | 2 - 3 | 309 | 3154405 | 95 | |
| BC098237.1 | - | 2 - 3 | 314 | 3825593 | 95 | |
| BF715142.1 | - | 2 - 3 | 305 | 95 | ||
More Information
Set Membership: |
|

Ap1s3
Genomic Map