Assay Details
| Gene Symbol: Ap1s3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183027.2 | NP_898848.1 | 3 - 4 | 417 | 128 | |
| GenBank mRNA | AK028013.1 | - | 3 - 4 | 402 | 128 | |
| BC054111.1 | - | 3 - 4 | 421 | 3154405 | 128 | |
| BC098237.1 | - | 3 - 4 | 426 | 3825593 | 128 | |
More Information
Set Membership: |
|

Ap1s3
Genomic Map