Assay Details
| Gene Symbol: Zfp691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183140.3 | NP_898963.2 | 1 - 2 | 817 | 99 | |
| GenBank mRNA | AK032431.1 | - | 1 - 2 | 819 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK032431.1, NM_183140.3 transcripts.
Set Membership: |
|

Zfp691
Genomic Map