Assay Details
| Gene Symbol: Marveld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183195.2 | NP_899018.1 | 1 - 2 | 2167 | 105 | |
| GenBank mRNA | AK132243.1 | - | 1 - 2 | 2158 | 105 | |
| BC044794.1 | - | 1 - 2 | 593 | 105 | ||
| BC054384.1 | - | 1 - 2 | 2130 | 6475446 | 105 | |
More Information
Set Membership: |
|

Marveld1
Genomic Map