Assay Details
| Gene Symbol: Ranbp3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198024.2 | NP_932141.1 | 1254 | 108 | ||
| GenBank mRNA | AK048152.1 | - | 1202 | 108 | ||
| BC058706.1 | - | 1084 | 6839130 | 108 | ||
More Information
Set Membership: |
|

Ranbp3l
Genomic Map