Assay Details
| Gene Symbol: Rai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198409.3 | NP_940801.2 | 1 - 2 | 309 | 78 | |
| GenBank mRNA | BC061184.1 | - | 1 - 2 | 235 | 30300326 | 78 |
| BY100054.1 | - | 1 - 2 | 333 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC061184.1, NM_198409.3 transcripts.
Set Membership: |
|

Rai2
Genomic Map