Assay Details
| Gene Symbol: Hn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198937.2 | NP_945175.1 | 1 - 2 | 57 | 119 | |
| GenBank mRNA | AK150226.1 | - | 1 - 2 | 69 | 119 | |
| AK152133.1 | - | 1 - 2 | 73 | 119 | ||
| AK152573.1 | - | 1 - 2 | 71 | 119 | ||
| AK166213.1 | - | 1 - 2 | 71 | 119 | ||
| AK167223.1 | - | 1 - 2 | 41 | 119 | ||
| AK169647.1 | - | 104 | 119 | |||
| BC057024.1 | - | 1 - 2 | 57 | 6815840 | 119 | |
| BC080740.1 | - | 1 - 2 | 97 | 30460749 | 119 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK167223.1 transcripts.
Set Membership: |
|

Hn1l
Genomic Map