Assay Details
| Gene Symbol: Prlhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201615.2 | NP_963909.2 | 1 | 1216 | 99 | |
| GenBank mRNA | BC125649.1 | - | 1 | 1148 | 40130164 | 99 |
| BC125651.1 | - | 1 | 1148 | 40130166 | 99 | |
More Information
Set Membership: |
|

Prlhr
Genomic Map