Assay Details
| Gene Symbol: Olfr845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207145.2 | NP_997028.1 | 1 | 417 | 80 | |
| GenBank mRNA | BC127982.1 | - | 1 | 418 | 40135240 | 80 |
More Information
Set Membership: |
|

Olfr845
Genomic Map