Assay Details
| Gene Symbol: Krt78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212487.4 | NP_997652.4 | 5 - 6 | 970 | 59 | |
| GenBank mRNA | AY651827.1 | - | 5 - 6 | 795 | 59 | |
| BC119600.1 | - | 5 - 6 | 907 | 59 | ||
| BC119601.1 | - | 5 - 6 | 907 | 59 | ||
| BC127241.1 | - | 5 - 6 | 907 | 59 | ||
| BC127618.1 | - | 5 - 6 | 907 | 59 | ||
More Information
Set Membership: |
|

Krt78
Genomic Map