Assay Details
| Gene Symbol: Tomt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081679.1 | NP_001075148.1 | 1 - 2 | 61 | 87 | |
| GenBank mRNA | DQ854743.1 | - | 1 - 2 | 61 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the DQ854743.1, NM_001081679.1 transcripts.
Set Membership: |
|

Tomt
Genomic Map