Assay Details
| Gene Symbol: Ngfrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110233.1 | NP_001103703.1 | 2 - 3 | 224 | 77 | |
| GenBank mRNA | AV574205.1 | - | 2 - 3 | 136 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001110233.1 transcripts.
Set Membership: |
|

Ngfrap1
Genomic Map