Assay Details
| Gene Symbol: Ngfrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110234.1 | NP_001103704.1 | 2 - 3 | 221 | 98 | |
| GenBank mRNA | AK003294.1 | - | 2 - 3 | 175 | 98 | |
| CB055608.1 | - | 2 - 3 | 120 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003294.1, NM_001110234.1 transcripts.
Set Membership: |
|

Ngfrap1
Genomic Map