Assay Details
| Gene Symbol: Pcdh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122758.1 | NP_001116230.1 | 2 - 3 | 4482 | 60 | |
| GenBank mRNA | AB011255.1 | - | 1 - 2 | 224 | 60 | |
| AK039737.1 | - | 2 - 3 | 804 | 60 | ||
| AY861419.1 | - | 2 - 3 | 692 | 60 | ||
More Information
Set Membership: |
|

Pcdh7
Genomic Map