Assay Details
| Gene Symbol: Tpgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142698.1 | NP_001136170.1 | 6 - 7 | 926 | 92 | |
| GenBank mRNA | AK141632.1 | - | 6 - 7 | 743 | 92 | |
More Information
Set Membership: |
|

Tpgs2
Genomic Map