Assay Details
| Gene Symbol: Rfx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145660.1 | NP_001139132.1 | 2 - 3 | 234 | 70 | |
| GenBank mRNA | AK133372.1 | - | 2 - 3 | 245 | 70 | |
More Information
Set Membership: |
|

Rfx8
Genomic Map