Assay Details
| Gene Symbol: Gpatch8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159492.1 | NP_001152964.1 | 5 - 6 | 377 | 95 | |
| GenBank mRNA | BC035255.1 | - | 2 - 3 | 386 | 95 | |
| CV555545.1 | - | 3 - 4 | 214 | 95 | ||
More Information
Set Membership: |
|

Gpatch8
Genomic Map