Assay Details
Gene Symbol: Tmem8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159602.1 | NP_001153074.1 | 4 - 5 | 512 | 66 | |
NM_025376.3 | NP_079652.1 | 4 - 5 | 664 | 66 | ||
GenBank mRNA | AK003298.1 | - | 4 - 5 | 666 | 66 | |
AK004477.1 | - | 4 - 5 | 632 | 66 | ||
BC062145.1 | - | 4 - 5 | 590 | 30286808 | 66 | |
CX564956.1 | - | 4 - 5 | 513 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
