Assay Details
| Gene Symbol: Trappc3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162937.1 | NP_001156409.1 | 2 - 3 | 267 | 80 | |
| GenBank mRNA | BC099502.1 | - | 2 - 3 | 267 | 30938327 | 80 |
More Information
Set Membership: |
|

Trappc3l
Genomic Map