Assay Details
| Gene Symbol: Trappc3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162937.1 | NP_001156409.1 | 3 - 4 | 368 | 109 | |
| GenBank mRNA | BC099502.1 | - | 3 - 4 | 368 | 30938327 | 109 |
More Information
Set Membership: |
|

Trappc3l
Genomic Map