Assay Details
| Gene Symbol: Otud6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163191.1 | NP_001156663.1 | 1 | 821 | 62 | |
| GenBank mRNA | BC153817.1 | - | 1 | 821 | 40050367 | 62 |
More Information
Set Membership: |
|

Otud6a
Genomic Map