Assay Details
| Gene Symbol: Smim10l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163445.2 | NP_001156917.1 | 1 | 2223 | 92 | |
| GenBank mRNA | AK077261.1 | - | 1 | 2202 | 92 | |
| AW546677.2 | - | 1 | 284 | 92 | ||
More Information
Set Membership: |
|

Smim10l1
Genomic Map