Assay Details
| Gene Symbol: Pom121l12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164166.1 | NP_001157638.1 | 1 | 326 | 101 | |
| GenBank mRNA | AK160151.1 | - | 1 | 292 | 101 | |
| BY716815.1 | - | 1 | 292 | 101 | ||
More Information
Set Membership: |
|

Pom121l12
Genomic Map