Assay Details
| Gene Symbol: AU022252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012400.2 | NP_001012400.1 | 4 - 5 | 483 | 76 | |
| GenBank mRNA | AK158349.1 | - | 4 - 5 | 441 | 76 | |
| AK167445.1 | - | 4 - 5 | 445 | 76 | ||
| BC035522.1 | - | 4 - 5 | 466 | 76 | ||
| BC089614.1 | - | 4 - 5 | 465 | 6748514 | 76 | |
More Information
Set Membership: |
|

AU022252
Genomic Map