Assay Details
| Gene Symbol: Scn4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013390.2 | NP_001013408.1 | 4 - 5 | 597 | 90 | |
| GenBank mRNA | AB118946.1 | - | 4 - 5 | 884 | 90 | |
| AY669079.1 | - | 4 - 5 | 597 | 90 | ||
| BC050045.1 | - | 1 - 2 | 136 | 90 | ||
| BC139399.1 | - | 4 - 5 | 676 | 8862421 | 90 | |
| BC139400.1 | - | 4 - 5 | 676 | 8862422 | 90 | |
More Information
Set Membership: |
|

Scn4b
Genomic Map