Assay Details
Gene Symbol: Syna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013751.2 | NP_001013773.1 | 1 | 1721 | 102 | |
GenBank mRNA | BC138919.1 | - | 1 | 1721 | 8861941 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
