Assay Details
| Gene Symbol: Aste1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164828.1 | NP_001158300.1 | 2 - 3 | 282 | 106 | |
| GenBank mRNA | AK006153.1 | - | 2 - 3 | 284 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001164828.1 transcripts.
Set Membership: |
|

Aste1
Genomic Map