Assay Details
| Gene Symbol: Tmem11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168507.1 | NP_001161979.1 | 2 - 3 | 615 | 75 | |
| GenBank mRNA | CX211768.1 | - | 2 - 3 | 349 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001168507.1 transcripts.
Set Membership: |
|

Tmem11
Genomic Map