Assay Details
| Gene Symbol: Cd1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289449.1 | NP_001276378.1 | 2 - 3 | 277 | 129 | |
| GenBank mRNA | BC057149.1 | - | 2 - 3 | 277 | 5328694 | 129 |
More Information
Set Membership: |
|

Cd1d2
Genomic Map