Assay Details
Gene Symbol: Ly6g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001310438.1 | NP_001297367.1 | 197 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001310438.1 transcripts.
Set Membership: |
![]() ![]() |
