Assay Details
| Gene Symbol: Klk1b8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008457.2 | NP_032483.1 | 3 - 4 | 515 | 111 | |
| GenBank mRNA | BC031798.1 | - | 3 - 4 | 520 | 4167600 | 111 |
| X03994.1 | - | 3 - 4 | 514 | 111 | ||
More Information
Set Membership: |
|

Klk1b8
Genomic Map