Assay Details
| Gene Symbol: Rpl26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009080.2 | NP_033106.1 | 1 - 2 | 64 | 91 | |
| GenBank mRNA | BY110756.1 | - | 1 - 2 | 65 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_009080.2 transcripts.
Set Membership: |
|

Rpl26
Genomic Map