Assay Details
| Gene Symbol: Clca3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF047838.1 | - | 575 | 127 | ||
| AF052746.1 | - | 4 - 5 | 604 | 127 | ||
| AK032511.1 | - | 4 - 5 | 574 | 127 | ||
| AK087213.1 | - | 4 - 5 | 576 | 127 | ||
| AK142211.1 | - | 4 - 5 | 576 | 127 | ||
| AK161805.1 | - | 4 - 5 | 577 | 127 | ||
| BC010260.1 | - | 4 - 5 | 813 | 3710379 | 127 | |
| BC132342.1 | - | 4 - 5 | 578 | 40130619 | 127 | |
| BC145057.1 | - | 578 | 9053605 | 127 | ||
More Information
Set Membership: |
|

Clca3a1
Genomic Map