Assay Details
Gene Symbol: Zfp235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019941.2 | NP_064325.2 | 3 - 4 | 512 | 91 | |
GenBank mRNA | AB221682.1 | - | 1 - 2 | 144 | 91 | |
BC099965.1 | - | 3 - 4 | 433 | 30084081 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
